Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282855.1 | NP_001269784.1 | 12, 13 | 2775 | ||
NM_001321293.1 | NP_001308222.1 | 12, 13 | 2787 | |||
NM_178827.4 | NP_849149.3 | 12, 13 | 2375 | |||
NR_104244.1 | 13, 14 | 2558 | ||||
NR_104245.1 | 11, 12 | 2199 | ||||
XM_005250161.3 | XP_005250218.1 | 13, 14 | 2490 | |||
XM_011515833.2 | XP_011514135.1 | 13, 14 | 2303 | |||
XM_011515834.2 | XP_011514136.1 | 12, 13 | 2608 | |||
XM_017011771.1 | XP_016867260.1 | 13, 14 | 2289 | |||
XM_017011772.1 | XP_016867261.1 | 7, 8 | 1951 | |||
GenBank mRNA | AK093153.1 | - | 12, 13 | 2295 | ||
BC026173.1 | - | 13, 14 | 2562 | 4838069 | ||
BC032838.2 | - | 12, 13 | 2300 | |||
BC036121.1 | - | 12, 13 | 2775 | 5296826 | ||
BC068607.1 | - | 12, 13 | 2300 | |||
BC091520.1 | - | 12, 13 | 2230 | 30379054 | ||
BC128181.1 | - | 12, 13 | 2274 | 40113310 | ||
BC128182.1 | - | 11, 12 | 2098 | 40113312 |