Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167961.2 | NP_001161433.1 | 1, 2 | 329 | ||
NM_153033.4 | NP_694578.1 | 1, 2 | 329 | |||
GenBank mRNA | AK056631.1 | - | 1, 2 | 252 | ||
BC042482.1 | - | 1, 2 | 267 | 5165722 | ||
DA493191.1 | - | 1, 2 | 332 | |||
DB095774.1 | - | 1, 2 | 307 |