Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152559.2 | NP_689772.2 | 5 | 490 | ||
XM_017011777.1 | XP_016867266.1 | 5 | 492 | |||
XM_017011778.1 | XP_016867267.1 | 5 | 614 | |||
GenBank mRNA | AF534110.1 | - | 5 | 501 | ||
AK301597.1 | - | 4 | 345 | |||
AY354928.1 | - | 5 | 500 | |||
BC030295.2 | - | 4 | 454 | 5396423 |