Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144962.2 | NP_659399.2 | 3 | 271 | ||
XM_011544414.2 | XP_011542716.1 | 3 | 326 | |||
GenBank mRNA | AY037148.1 | - | 3 | 244 | ||
AY359109.1 | - | 3 | 267 | |||
AY730275.1 | - | 3 | 222 | |||
BC020779.1 | - | 3 | 263 | 4700840 | ||
BF853000.1 | - | 3 | 271 | |||
FJ224332.1 | - | 3 | 263 |