Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317906.1 | NP_001304835.1 | 8 | 1699 | ||
NM_001317907.1 | NP_001304836.1 | 9 | 1616 | |||
NM_152562.3 | NP_689775.2 | 9 | 1616 | |||
XM_006716294.2 | XP_006716357.1 | 9 | 1631 | |||
XM_011544415.1 | XP_011542717.1 | 10 | 1703 | |||
XM_011544416.2 | XP_011542718.1 | 10 | 1397 | |||
XM_011544417.2 | XP_011542719.1 | 10 | 1454 | |||
XM_011544418.1 | XP_011542720.1 | 10 | 1667 | |||
GenBank mRNA | AK092164.1 | - | 4 | 233 | ||
AK292505.1 | - | 8 | 1431 | |||
AK299278.1 | - | 9 | 1322 | |||
AL833396.1 | - | 6 | 721 | |||
AL833627.1 | - | 8 | 1393 | |||
BC036214.1 | - | 6 | 744 | 5267276 | ||
BX649008.1 | - | 5 | 607 |