Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317906.1 | NP_001304835.1 | 2, 3 | 865 | ||
NM_001317907.1 | NP_001304836.1 | 3, 4 | 782 | |||
NM_152562.3 | NP_689775.2 | 3, 4 | 782 | |||
XM_011544418.1 | XP_011542720.1 | 3, 4 | 761 | |||
GenBank mRNA | AK292505.1 | - | 2, 3 | 597 | ||
AK299278.1 | - | 3, 4 | 488 | |||
AL833627.1 | - | 2, 3 | 559 | |||
BM802460.1 | - | 3, 4 | 451 |