Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194291.2 | NP_919267.2 | 6, 7 | 1155 | ||
XM_011516847.2 | XP_011515149.1 | 6, 7 | 1149 | |||
XM_017013104.1 | XP_016868593.1 | 4, 5 | 215 | |||
GenBank mRNA | BC017881.1 | - | 4, 5 | 259 | ||
BC032396.1 | - | 6, 7 | 456 | 5197382 | ||
BC041379.1 | - | 6, 7 | 1152 | 5275048 |