Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 8 | 815 | ||
NM_001168340.1 | NP_001161812.1 | 8 | 821 | |||
NM_001168341.1 | NP_001161813.1 | 6 | 614 | |||
NM_001168342.1 | NP_001161814.1 | 6 | 696 | |||
NM_152574.2 | NP_689787.2 | 8 | 821 | |||
XM_011517732.2 | XP_011516034.1 | 7 | 618 | |||
XM_017014310.1 | XP_016869799.1 | 10 | 1184 | |||
XM_017014311.1 | XP_016869800.1 | 9 | 1116 | |||
XM_017014312.1 | XP_016869801.1 | 7 | 686 | |||
GenBank mRNA | AK091187.1 | - | 8 | 707 | ||
AK093459.1 | - | 8 | 888 | |||
AK298581.1 | - | 6 | 469 | |||
AK298998.1 | - | 8 | 793 | |||
AK301427.1 | - | 6 | 612 | |||
AK310271.1 | - | 9 | 679 | |||
AK310482.1 | - | 8 | 821 | |||
BC042417.1 | - | 7 | 688 | |||
BC142985.1 | - | 8 | 794 | 8860428 |