Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286950.1 | NP_001273879.1 | 5 | 513 | ||
NM_001286951.1 | NP_001273880.1 | 5 | 557 | |||
NM_001286952.1 | NP_001273881.1 | 4 | 432 | |||
NM_001286953.1 | NP_001273882.1 | 4 | 421 | |||
NM_001286954.1 | NP_001273883.1 | 4, 5 | 509 | |||
NM_144964.3 | NP_659401.2 | 6 | 666 | |||
NR_104612.1 | 6 | 778 | ||||
XM_005251375.2 | XP_005251432.1 | 4 | 427 | |||
XM_005251379.2 | XP_005251436.1 | 6 | 697 | |||
XM_011517735.2 | XP_011516037.1 | 6 | 688 | |||
XM_011517736.2 | XP_011516038.1 | 6 | 693 | |||
XM_011517738.2 | XP_011516040.1 | 4 | 457 | |||
XM_011517739.2 | XP_011516041.1 | 4 | 463 | |||
XM_017014313.1 | XP_016869802.1 | 6 | 706 | |||
GenBank mRNA | AK056017.1 | - | 6 | 664 | ||
AK092718.1 | - | 5 | 547 | |||
AK294219.1 | - | 4 | 428 | |||
AK295733.1 | - | 5 | 503 | |||
AK304516.1 | - | 6 | 773 | |||
AK315922.1 | - | 4 | 411 | |||
BC012175.1 | - | 6 | 764 | 4645858 | ||
BC057774.1 | - | 6 | 653 | 30338757 | ||
CR986760.1 | - | 4, 5 | 496 |