Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 4 | 454 | ||
NM_144965.2 | NP_659402.1 | 4 | 493 | |||
XM_005251733.1 | XP_005251790.1 | 3 | 305 | |||
XM_005251734.1 | XP_005251791.1 | 4 | 478 | |||
XM_006716970.1 | XP_006717033.1 | 4 | 478 | |||
XM_006716971.1 | XP_006717034.1 | 3 | 305 | |||
XM_011518283.1 | XP_011516585.1 | 4 | 491 | |||
XM_011518285.1 | XP_011516587.1 | 4 | 491 | |||
XM_011518289.2 | XP_011516591.1 | 4 | 402 | |||
XM_011518291.1 | XP_011516593.1 | 4 | 478 | |||
XM_011518293.1 | XP_011516595.1 | 4 | 491 | |||
XM_011518294.2 | XP_011516596.1 | 4 | 491 | |||
XM_011518295.1 | XP_011516597.1 | 4 | 491 | |||
XM_011518297.1 | XP_011516599.1 | 4 | 491 | |||
XM_011518298.2 | XP_011516600.1 | 4 | 491 | |||
GenBank mRNA | AK057342.1 | - | 4 | 478 | ||
AK302422.1 | - | 3 | 305 | |||
AK302737.1 | - | 4 | 439 | |||
BC031281.1 | - | 4 | 491 | 5295890 |