Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 10, 11 | 1710 | ||
NM_144966.5 | NP_659403.4 | 34, 35 | 6829 | |||
XM_005251382.3 | XP_005251439.1 | 33, 34 | 7077 | |||
XM_005251384.4 | XP_005251441.1 | 12, 13 | 2123 | |||
XM_006716729.3 | XP_006716792.1 | 13, 14 | 2368 | |||
XM_011517758.2 | XP_011516060.1 | 10, 11 | 1759 | |||
XM_017014316.1 | XP_016869805.1 | 33, 34 | 7104 | |||
XM_017014317.1 | XP_016869806.1 | 34, 35 | 6647 | |||
XM_017014318.1 | XP_016869807.1 | 34, 35 | 7402 | |||
XM_017014319.1 | XP_016869808.1 | 35, 36 | 7306 | |||
XM_017014320.1 | XP_016869809.1 | 34, 35 | 7233 | |||
XM_017014321.1 | XP_016869810.1 | 33, 34 | 6896 | |||
XM_017014322.1 | XP_016869811.1 | 33, 34 | 6690 | |||
XM_017014323.1 | XP_016869812.1 | 33, 34 | 6363 | |||
XM_017014324.1 | XP_016869813.1 | 32, 33 | 6954 | |||
XM_017014325.1 | XP_016869814.1 | 33, 34 | 7104 | |||
XM_017014326.1 | XP_016869815.1 | 32, 33 | 5920 | |||
XM_017014327.1 | XP_016869816.1 | 29, 30 | 5289 | |||
GenBank mRNA | AB160987.1 | - | 32, 33 | 6013 | ||
AK058190.1 | - | 7, 8 | 864 | |||
BC031064.1 | - | 10, 11 | 1710 | |||
BX537574.1 | - | 27, 28 | 5404 | |||
BX641104.1 | - | 4, 5 | 6334 |