Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139246.4 | NP_640339.4 | 8 | 1487 | ||
XM_011518312.2 | XP_011516614.1 | 7 | 950 | |||
XM_017014342.1 | XP_016869831.1 | 7 | 1012 | |||
GenBank mRNA | AF258575.1 | - | 5 | 701 | ||
AK056453.1 | - | 6 | 614 | |||
AK056752.1 | - | 1 | 48 | |||
AK094043.1 | - | 8 | 1200 | |||
AK122809.1 | - | 6 | 1601 | |||
AK128615.1 | - | 9 | 1153 | |||
AK291199.1 | - | 8 | 1108 | |||
BC035604.1 | - | 8 | 1134 | 4385905 | ||
BX538308.1 | - | 8 | 1128 | |||
BX648493.1 | - | 8 | 1126 | |||
KU178877.1 | - | 7 | 918 | |||
KU178878.1 | - | 7 | 890 |