Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170779.1 | NP_001164250.1 | 4, 5 | 690 | ||
NM_138819.3 | NP_620174.1 | 4, 5 | 690 | |||
XM_005262382.1 | XP_005262439.1 | 5, 6 | 468 | |||
XM_005262383.2 | XP_005262440.1 | 5, 6 | 468 | |||
XM_005262384.4 | XP_005262441.1 | 5, 6 | 468 | |||
XM_005262386.3 | XP_005262443.1 | 5, 6 | 468 | |||
XM_006724734.3 | XP_006724797.1 | 5, 6 | 556 | |||
XM_006724735.1 | XP_006724798.1 | 5, 6 | 468 | |||
XM_006724736.2 | XP_006724799.1 | 5, 6 | 468 | |||
XM_011531295.1 | XP_011529597.1 | 5, 6 | 468 | |||
XM_011531296.2 | XP_011529598.1 | 5, 6 | 468 | |||
XM_011531297.2 | XP_011529599.1 | 5, 6 | 468 | |||
XM_011531298.2 | XP_011529600.1 | 5, 6 | 468 | |||
XM_011531299.1 | XP_011529601.1 | 2, 3 | 68 | |||
XM_011531300.2 | XP_011529602.1 | 2, 3 | 68 | |||
XM_011531301.2 | XP_011529603.1 | 2, 3 | 63 | |||
XM_011531302.2 | XP_011529604.1 | 2, 3 | 64 | |||
XM_017029308.1 | XP_016884797.1 | 5, 6 | 468 | |||
XM_017029309.1 | XP_016884798.1 | 2, 3 | 145 | |||
GenBank mRNA | AK130840.1 | - | 1, 2 | 26 | ||
AK295485.1 | - | 6, 7 | 1325 | |||
BC017868.1 | - | 4, 5 | 689 | 4699951 | ||
BC063491.1 | - | 4, 5 | 630 | 5229041 |