Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173525.2 | NP_775796.2 | 1 | 105 | ||
XM_011519926.2 | XP_011518228.1 | 1 | 2676 | |||
XM_011519927.2 | XP_011518229.1 | 1 | 2676 | |||
GenBank mRNA | BC031612.1 | - | 1 | 105 | 5171107 |