Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181783.3 | NP_861448.2 | 3 | 495 | ||
GenBank mRNA | AJ697717.1 | - | 3 | 365 | ||
AK074973.1 | - | 2 | 122 | |||
AK290436.1 | - | 3 | 495 | |||
AK302755.1 | - | 3 | 495 | |||
AK310375.1 | - | 3 | 495 | |||
AK310393.1 | - | 3 | 495 | |||
BC117175.1 | - | 2 | 316 | 40125726 | ||
BC117177.1 | - | 2 | 317 | 40125728 | ||
BX538169.1 | - | 3 | 468 | |||
CR749244.1 | - | 3 | 447 | |||
CR749309.1 | - | 3 | 481 | |||
CR933632.1 | - | 3 | 446 |