Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 13 | 1931 | ||
GenBank mRNA | AF536216.1 | - | 13 | 1904 | ||
AK075263.1 | - | 13 | 1589 | |||
AK313788.1 | - | 13 | 1900 | |||
AY081220.1 | - | 13 | 1905 | |||
BC110492.2 | - | 13 | 1566 | 40007567 |