Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPTC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139283.1 | NP_644812.1 | 6 | 1103 | ||
GenBank mRNA | AF385435.1 | - | 6 | 1103 | ||
AK124744.1 | - | 7 | 947 | |||
AY357944.1 | - | 6 | 872 | |||
BC111551.1 | - | 7 | 905 | 40009728 | ||
CR749216.1 | - | 3 | 225 |