Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 14, 15 | 1648 | ||
NM_001204505.2 | NP_001191434.1 | 11, 12 | 1269 | |||
NM_001204506.2 | NP_001191435.1 | 11, 12 | 1269 | |||
NM_001297429.1 | NP_001284358.1 | 9, 10 | 1036 | |||
NM_152910.5 | NP_690874.2 | 13, 14 | 1611 | |||
NM_178009.4 | NP_821077.1 | 13, 14 | 1611 | |||
NR_123714.1 | 11, 12 | 1269 | ||||
NR_123715.1 | 13, 14 | 1882 | ||||
GenBank mRNA | AB078967.1 | - | 13, 14 | 1566 | ||
AB078968.1 | - | 13, 14 | 1566 | |||
AK122909.1 | - | 11, 12 | 1234 | |||
AK289546.1 | - | 14, 15 | 1650 | |||
AK302631.1 | - | 9, 10 | 996 | |||
AK302727.1 | - | 11, 12 | 1269 | |||
BC043292.2 | - | 11, 12 | 1226 | 5297895 |