Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180989.5 | NP_851320.1 | 8, 9 | 1233 | ||
GenBank mRNA | AK126794.1 | - | 8, 9 | 1138 | ||
AK289850.1 | - | 8, 9 | 1480 | |||
AL832739.1 | - | 3, 4 | 2280 | |||
BC033906.1 | - | 7, 8 | 1027 | |||
BC052243.1 | - | 8, 9 | 1180 | 6083073 | ||
BG112926.1 | - | 182 | ||||
BI856811.1 | - | 6, 7 | 667 | |||
BX348762.1 | - | 3, 4 | 321 | |||
CA435391.1 | - | 1, 2 | 454 |