Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152592.3 | NP_689805.3 | 15 | 2658 | ||
XM_005267377.3 | XP_005267434.1 | 16 | 2755 | |||
XM_006720063.3 | XP_006720126.1 | 16 | 2855 | |||
XM_011536515.2 | XP_011534817.1 | 16 | 2790 | |||
XM_017021056.1 | XP_016876545.1 | 15 | 2642 | |||
GenBank mRNA | AK098471.1 | - | 10 | 1421 | ||
AK131436.1 | - | 16 | 2508 | |||
BC146604.1 | - | 16 | 2655 | 40123293 |