Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183387.2 | NP_899243.1 | 22 | 3431 | ||
XM_006720070.3 | XP_006720133.1 | 22 | 3231 | |||
XM_011536528.2 | XP_011534830.1 | 22 | 3232 | |||
XM_011536530.2 | XP_011534832.1 | 22 | 3231 | |||
XM_011536531.2 | XP_011534833.1 | 22 | 3231 | |||
XM_011536532.2 | XP_011534834.1 | 22 | 3231 | |||
XM_011536533.2 | XP_011534835.1 | 22 | 3117 | |||
XM_011536534.2 | XP_011534836.1 | 23 | 3149 | |||
XM_011536536.2 | XP_011534838.1 | 22 | 3231 | |||
XM_017021062.1 | XP_016876551.1 | 22 | 3231 | |||
XM_017021063.1 | XP_016876552.1 | 22 | 3231 | |||
XM_017021064.1 | XP_016876553.1 | 22 | 3117 | |||
XM_017021065.1 | XP_016876554.1 | 22 | 3231 | |||
XM_017021066.1 | XP_016876555.1 | 21 | 3032 | |||
XM_017021067.1 | XP_016876556.1 | 22 | 3552 | |||
XM_017021068.1 | XP_016876557.1 | 22 | 3231 | |||
XM_017021069.1 | XP_016876558.1 | 22 | 3476 | |||
XM_017021070.1 | XP_016876559.1 | 22 | 3231 | |||
GenBank mRNA | AK128086.1 | - | 15 | 1970 | ||
BC150639.1 | - | 22 | 3431 | 9057009 |