Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183387.2 | NP_899243.1 | 25 | 3804 | ||
XM_006720070.3 | XP_006720133.1 | 25 | 3604 | |||
XM_011536528.2 | XP_011534830.1 | 25 | 3605 | |||
XM_011536530.2 | XP_011534832.1 | 25 | 3604 | |||
XM_011536531.2 | XP_011534833.1 | 25 | 3604 | |||
XM_011536532.2 | XP_011534834.1 | 25 | 3604 | |||
XM_011536533.2 | XP_011534835.1 | 25 | 3490 | |||
XM_011536534.2 | XP_011534836.1 | 26 | 3522 | |||
XM_011536536.2 | XP_011534838.1 | 25 | 3604 | |||
XM_017021062.1 | XP_016876551.1 | 25 | 3604 | |||
XM_017021063.1 | XP_016876552.1 | 25 | 3604 | |||
XM_017021064.1 | XP_016876553.1 | 25 | 3490 | |||
XM_017021065.1 | XP_016876554.1 | 25 | 3604 | |||
XM_017021066.1 | XP_016876555.1 | 24 | 3405 | |||
XM_017021067.1 | XP_016876556.1 | 25 | 3925 | |||
XM_017021068.1 | XP_016876557.1 | 25 | 3604 | |||
XM_017021069.1 | XP_016876558.1 | 25 | 3849 | |||
XM_017021070.1 | XP_016876559.1 | 25 | 3604 | |||
GenBank mRNA | AK127024.1 | - | 4 | 298 | ||
AK128086.1 | - | 18 | 2343 | |||
BC150639.1 | - | 25 | 3804 | 9057009 |