Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173528.2 | NP_775799.2 | 2, 3 | 193 | ||
XM_006720408.2 | XP_006720471.1 | 3, 4 | 346 | |||
XM_011521280.1 | XP_011519582.1 | 2, 3 | 187 | |||
XM_017021963.1 | XP_016877452.1 | 3, 4 | 271 | |||
GenBank mRNA | AK095934.1 | - | 3, 4 | 346 | ||
AK302120.1 | - | 2, 3 | 231 | |||
AK307565.1 | - | 2, 3 | 188 | |||
BC062471.1 | - | 2, 3 | 193 | 30390735 |