Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286434.1 | NP_001273363.1 | 4 | 432 | ||
NM_153356.2 | NP_699187.1 | 5 | 540 | |||
XM_006720409.3 | XP_006720472.1 | 5 | 570 | |||
XM_006720410.3 | XP_006720473.1 | 4 | 462 | |||
XM_006720411.3 | XP_006720474.1 | 3 | 358 | |||
XM_011521281.2 | XP_011519583.1 | 5 | 569 | |||
XM_011521282.2 | XP_011519584.1 | 5 | 569 | |||
XM_011521283.2 | XP_011519585.1 | 5 | 569 | |||
XM_011521284.2 | XP_011519586.1 | 4 | 349 | |||
GenBank mRNA | AB449907.1 | - | 4 | 308 | ||
BC033516.1 | - | 5 | 499 | 5170475 | ||
HY018107.1 | - | 3 | 329 |