Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144975.3 | NP_659412.3 | 5 | 2350 | ||
XM_005257934.3 | XP_005257991.1 | 4 | 1670 | |||
GenBank mRNA | AK054668.1 | - | 5 | 2345 | ||
AK303299.1 | - | 5 | 2345 | |||
AK303632.1 | - | 4 | 1626 | |||
AL832814.1 | - | 2 | 530 | |||
BC125200.1 | - | 4 | 2286 | 40123369 | ||
BC125201.1 | - | 4 | 2286 | 40123372 | ||
BX647942.1 | - | 5 | 2346 |