Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138328.2 | NP_612201.1 | 9 | 1151 | ||
XM_006721733.3 | XP_006721796.1 | 8 | 1128 | |||
XM_006721734.3 | XP_006721797.1 | 8 | 1520 | |||
XM_011524443.2 | XP_011522745.1 | 7 | 969 | |||
XM_017024272.1 | XP_016879761.1 | 8 | 1055 | |||
XM_017024273.1 | XP_016879762.1 | 7 | 1030 | |||
XM_017024274.1 | XP_016879763.1 | 7 | 1358 | |||
XM_017024275.1 | XP_016879764.1 | 7 | 896 | |||
XM_017024276.1 | XP_016879765.1 | 6 | 870 | |||
XM_017024277.1 | XP_016879766.1 | 7 | 959 | |||
XM_017024278.1 | XP_016879767.1 | 7 | 986 | |||
XM_017024279.1 | XP_016879768.1 | 7 | 1794 | |||
XM_017024280.1 | XP_016879769.1 | 5 | 1010 | |||
GenBank mRNA | AJ313480.1 | - | 9 | 1151 | ||
AK124570.1 | - | 7 | 965 | |||
AK127490.1 | - | 3 | 310 | |||
BC100975.1 | - | 9 | 1150 | 40005244 | ||
BC100976.1 | - | 8 | 1050 | |||
BC100977.1 | - | 8 | 1127 | 40005246 | ||
BC100978.1 | - | 9 | 1130 | 40005247 |