Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172225.2 | NP_001165696.1 | 5 | 1940 | ||
NM_001172226.2 | NP_001165697.1 | 4 | 1844 | |||
NM_152606.4 | NP_689819.1 | 5 | 1969 | |||
GenBank mRNA | AB074186.1 | - | 1 | 651 | ||
AK122597.1 | - | 5 | 1890 | |||
AK131388.1 | - | 1 | 1894 | |||
AK290486.1 | - | 5 | 1890 | |||
AL832100.1 | - | 4 | 1845 | |||
AL832315.1 | - | 5 | 1934 | |||
AL832687.1 | - | 5 | 1893 | |||
BC126478.1 | - | 5 | 1740 | 8992194 | ||
BC126480.1 | - | 5 | 1740 | 8992196 | ||
BX537980.1 | - | 5 | 1885 |