Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256404.1 | NP_001243333.1 | 10 | 2142 | ||
NM_198459.3 | NP_940861.3 | 7 | 1602 | |||
GenBank mRNA | BX647950.1 | - | 7 | 1603 | ||
BX649075.1 | - | 7 | 728 | |||
CR749576.1 | - | 10 | 2066 |