Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256404.1 | NP_001243333.1 | 16 | 2933 | ||
NM_198459.3 | NP_940861.3 | 13 | 2393 | |||
GenBank mRNA | BX647950.1 | - | 13 | 2394 | ||
BX649075.1 | - | 13 | 1519 | |||
CR749576.1 | - | 16 | 2857 |