Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256404.1 | NP_001243333.1 | 20 | 3396 | ||
NM_198459.3 | NP_940861.3 | 17 | 2856 | |||
GenBank mRNA | AK124287.1 | - | 3 | 301 | ||
BX647950.1 | - | 17 | 2857 | |||
BX649075.1 | - | 18 | 2278 | |||
CR749576.1 | - | 20 | 3320 |