Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178422.5 | NP_848509.1 | 2 | 838 | ||
XM_005245745.3 | XP_005245802.1 | 3 | 660 | |||
XM_005245746.3 | XP_005245803.1 | 2 | 576 | |||
XM_011540861.1 | XP_011539163.1 | 3 | 483 | |||
XM_011540862.2 | XP_011539164.1 | 4 | 749 | |||
GenBank mRNA | AF313620.1 | - | 2 | 267 | ||
AY424285.1 | - | 1 | 399 | |||
BC034015.1 | - | 2 | 838 | 4400504 |
Set Membership: |
G-protein coupled receptor |