Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178821.1 | NP_849143.1 | 3 | 313 | ||
XM_011510755.1 | XP_011509057.1 | 4 | 922 | |||
XM_017003490.1 | XP_016858979.1 | 2 | 328 | |||
GenBank mRNA | AK097409.1 | - | 3 | 313 | ||
AK098821.1 | - | 3 | 261 | |||
AK127882.1 | - | 3 | 870 | |||
AK301861.1 | - | 4 | 313 | |||
BC036377.2 | - | 3 | 293 | 4824989 |