Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC138 SIRNA Assays ›
Gene Symbol:
CCDC138
Gene Name:
coiled-coil domain containing 138
Gene Aliases:
-
Chromosome Location:
Chr.2: 108786730 - 108885485 on Build GRCh38
UniGene ID:
Hs.362702
Species:
Human
siRNA ID
HSS136448
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC138
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303105.1
NP_001290034.1
3
255
NM_001303106.1
NP_001290035.1
3
255
NM_144978.2
NP_659415.1
3
255
NR_130121.1
3
255
XM_006712344.2
XP_006712407.1
3
240
XM_006712345.3
XP_006712408.1
3
240
XM_011510757.2
XP_011509059.1
3
240
XM_011510758.2
XP_011509060.1
2
217
XM_011510760.2
XP_011509062.1
3
240
XM_011510761.2
XP_011509063.1
3
240
XM_011510762.2
XP_011509064.1
3
240
XM_017003492.1
XP_016858981.1
3
253
XM_017003493.1
XP_016858982.1
2
213
XM_017003494.1
XP_016858983.1
3
253
XM_017003495.1
XP_016858984.1
3
253
XM_017003496.1
XP_016858985.1
3
240
XM_017003497.1
XP_016858986.1
3
253
XM_017003498.1
XP_016858987.1
3
240
XM_017003499.1
XP_016858988.1
3
253
XM_017003500.1
XP_016858989.1
2
212
XM_017003501.1
XP_016858990.1
3
149
XM_017003502.1
XP_016858991.1
3
205
XM_017003503.1
XP_016858992.1
3
144
XM_017003504.1
XP_016858993.1
2
159
GenBank mRNA
AK057307.1
-
3
242
AK125199.1
-
3
244
BC015847.1
-
3
236
BC038421.1
-
3
216
BC050579.1
-
3
248
5764613
DN919595.1
-
3
145
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline