Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM171B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177454.3 | NP_803237.3 | 8 | 1971 | ||
GenBank mRNA | AB075826.1 | - | 7 | 1420 | ||
AK091423.1 | - | 9 | 1876 | |||
AK094578.1 | - | 1 | 379 | |||
AK095604.1 | - | 5 | 1253 | |||
AK289737.1 | - | 8 | 1974 | |||
AL834264.1 | - | 1 | 373 | |||
BC060872.1 | - | 8 | 1977 | 30347998 |