Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113523.2 | NP_001106995.1 | 9 | 1325 | ||
NM_152615.2 | NP_689828.1 | 5 | 652 | |||
XM_005247160.3 | XP_005247217.1 | 8 | 2769 | |||
XM_011512475.2 | XP_011510777.1 | 8 | 2881 | |||
XM_011512476.2 | XP_011510778.1 | 8 | 2850 | |||
XM_011512477.2 | XP_011510779.1 | 7 | 2619 | |||
XM_011512478.2 | XP_011510780.1 | 9 | 1108 | |||
XM_011512479.2 | XP_011510781.1 | 7 | 583 | |||
XM_017005792.1 | XP_016861281.1 | 6 | 1750 | |||
GenBank mRNA | AK097515.1 | - | 11 | 1270 | ||
AK097916.1 | - | 5 | 589 | |||
BC101701.1 | - | 5 | 652 | 8069207 | ||
BC101703.1 | - | 5 | 652 | 8069209 | ||
DQ063586.1 | - | 9 | 1244 |