Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153330.4 | NP_699161.1 | 3 | 2081 | ||
XM_006713519.3 | XP_006713582.1 | 2 | 2978 | |||
GenBank mRNA | AI188219.1 | - | 1 | 393 | ||
AK125946.1 | - | 2 | 2978 | |||
AK126068.1 | - | 1 | 656 | |||
BC029521.1 | - | 1 | 674 | 5296554 | ||
BC050288.1 | - | 3 | 2081 | 5270636 |