Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168271.1 | NP_001161743.1 | 9 | 1474 | ||
NM_153002.2 | NP_694547.2 | 9 | 1486 | |||
XM_005247165.2 | XP_005247222.1 | 10 | 1763 | |||
XM_011512484.2 | XP_011510786.1 | 11 | 1836 | |||
XM_011512487.1 | XP_011510789.1 | 9 | 1388 | |||
XM_011512488.2 | XP_011510790.1 | 2 | 455 | |||
XM_017005795.1 | XP_016861284.1 | 10 | 1799 | |||
XM_017005796.1 | XP_016861285.1 | 10 | 1808 | |||
XM_017005797.1 | XP_016861286.1 | 10 | 1751 | |||
GenBank mRNA | AB209362.1 | - | 4 | 650 | ||
AF488739.1 | - | 9 | 1289 | |||
BC113701.1 | - | 9 | 1309 | 8322753 | ||
BC143606.1 | - | 9 | 1297 | 9052119 |