Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198621.2 | NP_001185550.1 | 3 | 5089 | ||
NM_194293.2 | NP_919269.2 | 2 | 5506 | |||
XM_005264909.3 | XP_005264966.1 | 2 | 5465 | |||
GenBank mRNA | AJ271461.1 | - | 1 | 634 | ||
AJ626899.1 | - | 3 | 5087 | |||
AJ626900.1 | - | 2 | 5514 | |||
AK095941.1 | - | 1 | 1129 | |||
AK096421.1 | - | 2 | 1651 | |||
AK122723.1 | - | 2 | 2486 | |||
AK126299.1 | - | 1 | 3956 | |||
AL713648.1 | - | 1 | 3594 | |||
AL831956.1 | - | 2 | 2874 | |||
AL832020.1 | - | 3 | 3125 | |||
AL832459.1 | - | 2 | 3167 | |||
AY375160.1 | - | 2 | 5506 | |||
BC127119.1 | - | 2 | 1637 | |||
BC139782.1 | - | 2 | 1637 | |||
CR749430.1 | - | 3 | 5089 |