Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152617.3 | NP_689830.2 | 6 | 2016 | ||
GenBank mRNA | AK093113.1 | - | 6 | 1402 | ||
BC017442.1 | - | 5 | 1848 | |||
BC033791.1 | - | 6 | 1959 | 5163887 |