Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136257.1 | NP_001129729.1 | 4 | 1004 | ||
NM_001136258.1 | NP_001129730.1 | 5 | 1196 | |||
NM_152621.5 | NP_689834.1 | 4 | 1280 | |||
XM_011531698.2 | XP_011530000.1 | 5 | 1334 | |||
XM_011531699.2 | XP_011530001.1 | 6 | 1208 | |||
XM_011531700.2 | XP_011530002.1 | 5 | 1105 | |||
XM_011531701.2 | XP_011530003.1 | 6 | 2219 | |||
XM_011531702.2 | XP_011530004.1 | 5 | 2147 | |||
XM_017007839.1 | XP_016863328.1 | 5 | 1338 | |||
XM_017007840.1 | XP_016863329.1 | 6 | 1460 | |||
XM_017007841.1 | XP_016863330.1 | 5 | 1334 | |||
GenBank mRNA | AF452717.1 | - | 5 | 1099 | ||
AK290344.1 | - | 5 | 1198 | |||
AK314049.1 | - | 4 | 995 | |||
AL833599.1 | - | 3 | 250 | |||
BC028705.1 | - | 4 | 1005 | 4823252 | ||
BC041369.2 | - | 4 | 1273 | 5273650 | ||
BX649142.1 | - | 3 | 661 |