Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297598.1 | NP_001284527.1 | 6 | 506 | ||
NM_001297599.1 | NP_001284528.1 | 6 | 491 | |||
NM_152622.4 | NP_689835.3 | 6 | 491 | |||
XM_011543216.1 | XP_011541518.1 | 6 | 526 | |||
XM_011543217.1 | XP_011541519.1 | 6 | 526 | |||
XM_017009149.1 | XP_016864638.1 | 7 | 527 | |||
GenBank mRNA | AK093273.1 | - | 4 | 418 | ||
AK299285.1 | - | 5 | 466 | |||
BC041348.2 | - | 6 | 488 | 5272287 | ||
BX537798.1 | - | 6 | 476 | |||
BX640907.1 | - | 6 | 494 | |||
BX647359.1 | - | 6 | 485 | |||
BX648294.1 | - | 6 | 486 | |||
CR749246.1 | - | 7 | 526 | |||
CR749805.1 | - | 7 | 526 | |||
DB136129.1 | - | 6 | 495 |