Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114393.1 | NP_001107865.1 | 8, 9 | 1162 | ||
NM_001114394.1 | NP_001107866.1 | 7, 8 | 1437 | |||
NM_001297744.1 | NP_001284673.1 | 8, 9 | 1150 | |||
NM_001297745.1 | NP_001284674.1 | 7, 8 | 1437 | |||
NM_173797.3 | NP_776158.2 | 8, 9 | 1196 | |||
XM_011543231.2 | XP_011541533.1 | 7, 8 | 1441 | |||
XM_017009153.1 | XP_016864642.1 | 8, 9 | 1511 | |||
XM_017009154.1 | XP_016864643.1 | 9, 10 | 1588 | |||
XM_017009155.1 | XP_016864644.1 | 8, 9 | 1530 | |||
XM_017009156.1 | XP_016864645.1 | 9, 10 | 1820 | |||
XM_017009157.1 | XP_016864646.1 | 7, 8 | 1429 | |||
XM_017009158.1 | XP_016864647.1 | 8, 9 | 1499 | |||
XM_017009159.1 | XP_016864648.1 | 8, 9 | 1518 | |||
XM_017009160.1 | XP_016864649.1 | 9, 10 | 1099 | |||
XM_017009161.1 | XP_016864650.1 | 9, 10 | 1593 | |||
XM_017009162.1 | XP_016864651.1 | 9, 10 | 1169 | |||
GenBank mRNA | AK095818.1 | - | 8, 9 | 1043 | ||
AL833136.1 | - | 7, 8 | 1423 | |||
BC026061.1 | - | 9, 10 | 1195 | 3640197 | ||
BC047581.1 | - | 8, 9 | 1129 | 4824607 |