Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138295.3 | NP_612152.1 | 31 | 4957 | ||
XM_017011798.1 | XP_016867287.1 | 32 | 11362 | |||
GenBank mRNA | AB061683.1 | - | 31 | 4923 |