Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152888.2 | NP_690848.1 | 63 | 4934 | ||
XM_011516883.2 | XP_011515185.1 | 62 | 4952 | |||
XM_011516884.2 | XP_011515186.1 | 61 | 4913 | |||
XM_011516885.2 | XP_011515187.1 | 61 | 4898 | |||
XM_011516886.2 | XP_011515188.1 | 63 | 4864 | |||
XM_011516887.2 | XP_011515189.1 | 59 | 3832 | |||
XM_011516889.2 | XP_011515191.1 | 52 | 3073 | |||
XM_017013150.1 | XP_016868639.1 | 63 | 4825 | |||
XM_017013151.1 | XP_016868640.1 | 59 | 4790 | |||
XM_017013152.1 | XP_016868641.1 | 58 | 3822 | |||
GenBank mRNA | AF406780.1 | - | 63 | 4921 | ||
BC042075.1 | - | 37 | 2159 | |||
BC144535.1 | - | 51 | 3592 | 9053062 |