Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194294.2 | NP_919270.2 | 2 | 348 | ||
GenBank mRNA | AK128691.1 | - | 2 | 348 | ||
BC113496.1 | - | 2 | 127 | |||
BC113498.1 | - | 2 | 127 | |||
EF052681.1 | - | 1 | 67 |