Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042413.1 | NP_001035878.1 | 11 | 3436 | ||
NM_152629.3 | NP_689842.3 | 10 | 2452 | |||
XM_005251386.4 | XP_005251443.1 | 10 | 2995 | |||
XM_005251387.4 | XP_005251444.1 | 10 | 3273 | |||
XM_005251388.4 | XP_005251445.1 | 9 | 2787 | |||
XM_011517763.2 | XP_011516065.1 | 11 | 3115 | |||
XM_011517764.2 | XP_011516066.1 | 11 | 2945 | |||
XM_011517766.2 | XP_011516068.1 | 10 | 3524 | |||
XM_011517767.2 | XP_011516069.1 | 10 | 2918 | |||
XM_017014361.1 | XP_016869850.1 | 10 | 3223 | |||
GenBank mRNA | AB209404.1 | - | 8 | 1045 | ||
AK096318.1 | - | 8 | 1079 | |||
BC033899.2 | - | 10 | 2452 | 4828458 | ||
DQ438877.1 | - | 3317 | ||||
DQ438878.1 | - | 8 | 1380 | |||
KU178885.1 | - | 9 | 2258 | |||
KU178887.1 | - | 6 | 707 |