Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWWP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098637.1 | NP_001092107.1 | 2 | 790 | ||
NM_138499.3 | NP_612508.3 | 2 | 790 | |||
XM_006717659.3 | XP_006717722.1 | 2 | 792 | |||
XM_011539387.2 | XP_011537689.1 | 2 | 792 | |||
GenBank mRNA | AF318365.1 | - | 1 | 441 | ||
AK128663.1 | - | 1 | 627 | |||
BC011630.2 | - | 1 | 262 | |||
BC068574.1 | - | 2 | 788 | 30344364 |