Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009998.3 | NP_001009998.1 | 1, 2 | 376 | ||
NM_032627.4 | NP_116016.1 | 1, 2 | 376 | |||
XM_006722666.2 | XP_006722729.1 | 1, 2 | 381 | |||
XM_006722668.2 | XP_006722731.1 | 1, 2 | 381 | |||
XM_017026433.1 | XP_016881922.1 | 1, 2 | 381 | |||
XM_017026436.1 | XP_016881925.1 | 1, 2 | 381 | |||
GenBank mRNA | AK098429.1 | - | 1, 2 | 282 | ||
BC000274.1 | - | 1, 2 | 283 | 3356283 | ||
BM851755.1 | - | 1, 2 | 386 | |||
BQ068310.1 | - | 415 | ||||
BU838485.1 | - | 1, 2 | 147 | |||
CX873722.1 | - | 1, 2 | 376 |