Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114397.1 | NP_001107869.1 | 21 | 2468 | ||
NM_020865.2 | NP_065916.2 | 21 | 2510 | |||
GenBank mRNA | AB040921.1 | - | 19 | 1962 | ||
AF217190.1 | - | 21 | 2502 | |||
AJ577133.1 | - | 21 | 2429 | |||
AJ577134.1 | - | 21 | 2387 | |||
AK096808.1 | - | 1 | 41 | |||
AK303720.1 | - | 2390 | ||||
AK314435.1 | - | 21 | 2510 | |||
BC036035.1 | - | 20 | 2387 | 5273384 |