Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 7, 8 | 1176 | ||
XM_005254872.3 | XP_005254929.1 | 7, 8 | 2092 | |||
XM_011521312.2 | XP_011519614.1 | 7, 8 | 2092 | |||
XM_017021984.1 | XP_016877473.1 | 6, 7 | 751 | |||
GenBank mRNA | AJ315735.1 | - | 7, 8 | 1093 | ||
AK131344.1 | - | 6, 7 | 722 |